Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-57]
USBIC2254-56D1
- Antibody type:Primary
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:MEM-57
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, BSA, 15mM sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MEM-57
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human