Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD3 Rabbit Monoclonal Antibody [clone: MRQ-39]
Anti-CD3 Rabbit Monoclonal Antibody [clone: MRQ-39]
Catalog # USBIC2254-02C
Supplier:  US Biological
Anti-CD3 Rabbit Monoclonal Antibody [clone: MRQ-39]
Catalog # USBIC2254-02C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3
  • Clonality:
    Monoclonal
  • Clone:
    MRQ-39
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, protebase, sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MRQ-39
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human