Anti-CTSH Mouse Polyclonal Antibody
USBIC2097-61E
- Antibody type:Primary
- Antigen name:cathepsin H
- Antigen symbol:CTSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: CTSH (cathepsin H)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype:
Reactivity: Human