Anti-CTSH Chicken Polyclonal Antibody
Catalog # USBIC2097-61C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cathepsin H
- Antigen symbol:CTSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:31 to 113
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: CTSH (cathepsin H)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human