Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CASP10 Rabbit polyclonal antibody
Anti-CASP10 Rabbit polyclonal antibody
Catalog # USBIC2088-50D
Supplier:  US Biological
Anti-CASP10 Rabbit polyclonal antibody
Catalog # USBIC2088-50D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    caspase 10, apoptosis-related cysteine peptidase
  • Antigen symbol:
    CASP10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Serum
  • Form:
    liquid
  • Amino acid number:
    373 to 392
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: CASP10 (caspase 10, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human