Anti-CFLAR Rabbit polyclonal antibody
Catalog # USBIC2087-42U
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CASP8 and FADD-like apoptosis regulator
- Antigen symbol:CFLAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:191 to 209
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CFLAR (CASP8 and FADD-like apoptosis regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human