Anti-S100A8 Mouse Monoclonal Antibody [clone: CF-145]
Catalog # USBIC1035-61E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A8
- Antigen symbol:S100A8
- Clonality:Monoclonal
- Clone:CF-145
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: S100A8 (S100 calcium binding protein A8)
Clonality: Monoclonal
Clone: CF-145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human