Anti-Bu-1a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K93]
Catalog # USBIB2788-50C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:BU-1A
- Clonality:Monoclonal
- Clone:5K93
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Bu-1a
Clonality: monoclonal
Clone: 5K93
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken