Anti-BNP Mouse monoclonal antibody unconjugated [clone: 27-4-3]
USBIB2702-34B-100
- Antibody type:Primary
- Antigen name:Brain Natriuretic Peptide
- Antigen symbol:BNP
- Clonality:Monoclonal
- Clone:27-4-3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:1 to 21
- Storage buffer:200mM Na Citrate-Tris, 500mM sodium chloride, pH 7,5, 0,02% sodium azide
- Sequence:HPLGSPGSASDLETSGLQEQR
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: monoclonal
Clone: 27-4-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human