Anti-BCL2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 7H161]
USBIB0807-06G
- Antibody type:Primary
- Antigen name:B-cell CLL/lymphoma 2
- Antigen symbol:BCL2
- Clonality:Monoclonal
- Clone:7H161
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Amino acid number:41 to 54
- Storage buffer:PBS, pH 7,2, 5% glycerol, 0,2% BSA, 0,04% sodium azide.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: monoclonal
Clone: 7H161
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human