Anti-NPPA Mouse Monoclonal Antibody [clone: A137-M6-19-B6]
USBIA4150-03G-100
- Antibody type:Primary
- Antigen name:Natriuretic peptide A
- Antigen symbol:NPPA
- Clonality:Monoclonal
- Clone:A137-M6-19-B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Form:lyophilized powder
- Amino acid number:40 to 55
- Sequence:CEPNEEAGAALSPLPEV
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: monoclonal
Clone: A137-M6-19-B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: