Anti-ATP12A Rabbit polyclonal antibody unconjugated
USBIA3999-87
- Antibody type:Primary
- Antigen name:ATPase, H+/K+ transporting, nongastric, alpha Polypeptide
- Antigen symbol:ATP12A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:839 to 868
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Flow Cytometry: 1:10:1:50; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500
Type: Primary
Antigen: ATP12A (ATPase, H+/K+ transporting, nongastric, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human