Anti-ATG4B Rabbit polyclonal antibody unconjugated
Catalog # USBIA3935-06M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:autophagy related 4B, cysteine peptidase
- Antigen symbol:ATG4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:350 to 400
- Storage buffer:PBS, 0,2% gelatin, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: ATG4B (autophagy related 4B, cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat