Anti-APAF1 Mouse Monoclonal Antibody [clone: 6G51]
Catalog # USBIA2298-64M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Apoptotic peptidase activating factor 1
- Antigen symbol:APAF1
- Clonality:Monoclonal
- Clone:6G51
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Antigen synonyms:CED4
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5-10 ?g/ml
Type: Primary
Antigen: APAF1 (apoptotic peptidase activating factor 1)
Clonality: monoclonal
Clone: 6G51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat