Anti-ADAMTSL1 Mouse Monoclonal Antibody [clone: 9H43]
USBIA0859-62A3
- Antibody type:Primary
- Antigen name:ADAMTS-like 1
- Antigen symbol:ADAMTSL1
- Clonality:Monoclonal
- Clone:9H43
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:29 to 525
- Storage buffer:0,2um sterile-filtered solution
- Storage temperature:Cool
- Concentration:~500 ug/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1-2?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: ADAMTSL1 (ADAMTS-like 1)
Clonality: monoclonal
Clone: 9H43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human