Anti-ADAMTSL1 Goat Polyclonal Antibody
Katalogové číslo USBIA0859-62A2
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ADAMTS-like 1
- Antigen symbol:ADAMTSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:29 to 525
- Storage buffer:0,2um sterile-filtered solution
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: ADAMTSL1 (ADAMTS-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human