Anti-ADAM23 Mouse Monoclonal Antibody [clone: 9H37]
Catalog # USBIA0859-41D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 23
- Antigen symbol:ADAM23
- Clonality:Monoclonal
- Clone:9H37
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:60 to 585
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 25 ?g/ml
Type: Primary
Antigen: ADAM23 (ADAM metallopeptidase domain 23)
Clonality: monoclonal
Clone: 9H37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human