Anti-ACVR2A Rabbit Polyclonal Antibody
USBIA0855-98B
- Antibody type:Primary
- Antigen name:activin A receptor, type IIA
- Antigen symbol:ACVR2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µg
- Pk:50 µG
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: ACVR2A (activin A receptor, type IIA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human