Anti-POMC Mouse Monoclonal Antibody [clone: 1.B.55]
Catalog # USBIA0758-07-500
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:proopiomelanocortin
- Antigen symbol:POMC
- Clonality:Monoclonal
- Clone:1.B.55
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: monoclonal
Clone: 1.B.55
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat