Anti-POMC Mouse Monoclonal Antibody [clone: 57]
Catalog # USBIA0758-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:proopiomelanocortin
- Antigen symbol:POMC
- Clonality:Monoclonal
- Clone:57
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Recommended dilution: Immunochemistry: 1:100-1:200
Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: monoclonal
Clone: 57
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: