Anti-ADIPOQ Mouse Monoclonal Antibody (Biotin) [clone: 9H11]
Catalog # USBIA0725-08E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Adiponectin
- Antigen symbol:ADIPOQ
- Clonality:Monoclonal
- Clone:9H11
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:19 to 244
- Storage buffer:PBS, 50ug BSA per 1ug antibody.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-2.0 ?g/ml
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: monoclonal
Clone: 9H11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human