Anti-N-Acetylglucosamine, O-linked Mouse Monoclonal Antibody [clone: 6D92]
USBIA0528-19
- Antibody type:Primary
- Antigen name:N-Acetylglucosamine, O-linked
- Clonality:Monoclonal
- Clone:6D92
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~2 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: N-Acetylglucosamine, O-linked
Clonality: monoclonal
Clone: 6D92
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: