Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
USBI0014-24D
- Antibody type:Primary
- Antigen symbol:14-3-3 delta/zeta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human