Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
Katalogové číslo USBI0014-24B
Dodavatel: US Biological
Specifikace
- Antibody type:Primary
- Antigen symbol:14-3-3 delta/zeta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifikace
O tomto produktu
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human