Order Entry
Czech Republic
ContactUsLinkComponent
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Catalog # USBI0014-04B
Supplier:  US Biological
Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Catalog # USBI0014-04B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 epsilon
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rabbit,
    Sheep,
    Chicken,
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Format:
    Serum
  • Form:
    liquid
  • Sequence:
    CGEEQNKEALQDVEDENQ
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: 14-3-3 epsilon (14-3-3 epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Rabbit, Rat, Sheep