Anti-14-3-3 beta Rabbit Polyclonal Antibody
USBI0014-02P1
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 141
- Storage buffer:0,1M Tris-glycine, pH 7, 0,01% thimerosal,10% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Recommended dilution: Western Blot: 1:5000-1:20000; Immunochemistry: 5 ?g/ml; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human