Anti-14-3-3 beta Mouse Monoclonal Antibody [clone: J2E9]
USBI0014-02N
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Monoclonal
- Clone:J2E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 246
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: Monoclonal
Clone: J2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human