Anti-14-3-3 beta Rabbit Polyclonal Antibody
USBI0014-02M
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:10mM PBS, pH 7,4, BSA, sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human