Anti-14-3-3 beta Rabbit Polyclonal Antibody
USBI0014-02H
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit,Sheep,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Format:Serum
- Form:liquid
- Sequence:AcTMDKSELVC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rabbit, Rat, Sheep