Anti-Pannexin-2 Mouse Monoclonal Antibody [clone: S121A-31]
Catalog # USBI167602
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Pannexin 2
- Antigen symbol:PANX2
- Clonality:Monoclonal
- Clone:S121A-31
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: Pannexin-2
Clonality: monoclonal
Clone: S121A-31
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat