Anti-SLC12A5 Mouse Monoclonal Antibody [clone: S1-12]
Catalog # USBI167594
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 12 (potassium/chloride transporter), member 5
- Antigen symbol:Solute carrier family 12, member 5
- Clonality:Monoclonal
- Clone:S1-12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 1:300
Type: Primary
Antigen: SLC12A5 (solute carrier family 12 (potassium/chloride transporter), member 5)
Clonality: monoclonal
Clone: S1-12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat