Anti-KCNIP3 Goat Polyclonal Antibody
USBI167385
- Antibody type:Primary
- Antigen name:Kv channel interacting Protein 3, calsenilin
- Antigen symbol:KCNIP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-HTPLSKKEGIK
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: KCNIP3 (Kv channel interacting protein 3, calsenilin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human