Anti-ARSH Goat Polyclonal Antibody
USBI151360
- Antibody type:Primary
- Antigen name:Arylsulfatase family, member H
- Antigen symbol:ARSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:4000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: ARSH (arylsulfatase family, member H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human