Anti-TSC2 Rabbit Polyclonal Antibody
USBI146057
- Antibody type:Primary
- Antigen name:tuberous sclerosis 2
- Antigen symbol:TSC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse