Anti-CASP1 Rabbit Polyclonal Antibody
Katalogové číslo USBI146037
Dodavatel: US Biological
Specifikace
- Antibody type:Primary
- Antigen name:caspase 1, apoptosis-related cysteine peptidase
- Antigen symbol:CASP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CASP1 (caspase 1, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human