Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ERMAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 767505]
Anti-ERMAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 767505]
Catalog # USBI145459
Supplier:  US Biological
Anti-ERMAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 767505]
Catalog # USBI145459
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    erythroblast membrane-associated Protein (Scianna blood group)
  • Antigen symbol:
    ERMAP
  • Clonality:
    Monoclonal
  • Clone:
    767505
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,5% BSA, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: ERMAP (erythroblast membrane-associated protein (Scianna blood group))
Clonality: monoclonal
Clone: 767505
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human