Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SYP Mouse Monoclonal Antibody [clone: Syn-58]
Anti-SYP Mouse Monoclonal Antibody [clone: Syn-58]
Catalog # USBI145243
Supplier:  US Biological
Anti-SYP Mouse Monoclonal Antibody [clone: Syn-58]
Catalog # USBI145243
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptophysin
  • Antigen symbol:
    SYP
  • Clonality:
    Monoclonal
  • Clone:
    Syn-58
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: SYP (synaptophysin)
Clonality: monoclonal
Clone: Syn-58
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat