Anti-Myosin, Skeletal, Slow Mouse Monoclonal Antibody [clone: IML-64]
USBI144941
- Antibody type:Primary
- Antigen name:Myosin, Skeletal, Slow
- Clonality:Monoclonal
- Clone:IML-64
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Myosin, Skeletal, Slow
Clonality: monoclonal
Clone: IML-64
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit, Rat