Anti-Myosin, Skeletal, Fast Mouse Monoclonal Antibody [clone: MY-38]
Catalog # USBI144940
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Myosin, Skeletal, Fast
- Clonality:Monoclonal
- Clone:MY-38
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Myosin, Skeletal, Fast
Clonality: monoclonal
Clone: MY-38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit, Rat