Anti-COL3 Mouse Monoclonal Antibody [clone: Col-29]
USBI144378
- Antibody type:Primary
- Antigen name:collagen, type III
- Antigen symbol:COL3
- Clonality:Monoclonal
- Clone:Col-29
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: monoclonal
Clone: Col-29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat