Anti-RNF38 Rabbit Polyclonal Antibody
USBI138297
- Antibody type:Primary
- Antigen name:Ring Finger Protein 38
- Antigen symbol:RNF38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: RNF38 (ring finger protein 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human