Anti-Interferon alpha 2a Rabbit Polyclonal Antibody
Catalog # USBI138054
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon alpha 2a
- Antigen symbol:IFNA2a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1500
Type: Primary
Antigen: Interferon alpha 2a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human