Anti-HLA Class 2 Antigen DR Mouse Monoclonal Antibody [clone: 13H268]
Catalog # USBI138019
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:HLA Class 2 Antigen DR
- Clonality:Monoclonal
- Clone:13H268
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Form:liquid
- Storage buffer:20mMTRIS, pH 8.0, 10mg BSA, 150mM sodium chloride.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:200 Tests
- Pk:200 Tests
Specifications
About this item
Type: Primary
Antigen: HLA Class 2 Antigen DR
Clonality: monoclonal
Clone: 13H268
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: