Anti-APC Rat Monoclonal Antibody [clone: 13H8]
Catalog # USBI137553
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:adenomatous polyposis coli
- Antigen symbol:APC
- Clonality:Monoclonal
- Clone:13H8
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: APC (adenomatous polyposis coli)
Clonality: monoclonal
Clone: 13H8
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse