Anti-Adenylate Cyclase Type 3 Rabbit Polyclonal Antibody
USBI136860
- Antibody type:Primary
- Antigen name:adenylate cyclase Type 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Rat,Mouse,Cat
- Western blot:Yes
- Form:liquid
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Immunofluorescence: 1:500-1:1000; Western Blot: 1:10000
Type: Primary
Antigen: Adenylate Cyclase Type 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat, Chicken, Human, Mouse, Rat