Anti-KDELR1 Mouse Monoclonal Antibody [clone: 2E7]
Catalog # USBI136785
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum Protein retention receptor 1
- Antigen symbol:KDELR1
- Clonality:Monoclonal
- Clone:2E7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Yeast,Drosophila,Xenopus laevis
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:50-1:500; Western Blot: 1:100-1:1000
Type: Primary
Antigen: KDELR1 (KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1)
Clonality: monoclonal
Clone: 20000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Drosophila, Xenopus laevis, Yeast