Anti-RPL17 Rabbit Polyclonal Antibody
Catalog # USBI136391
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein L17
- Antigen symbol:RPL17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Storage buffer:PBS,(without Mg2+, Ca2+), pH 7,4, 150mM sodium chloride, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: RPL17 (ribosomal protein L17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat