Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Prostaglandin Dehydrogenase 15 Rabbit Polyclonal Antibody
Anti-Prostaglandin Dehydrogenase 15 Rabbit Polyclonal Antibody
Catalog # USBI136350
Supplier:  US Biological
Anti-Prostaglandin Dehydrogenase 15 Rabbit Polyclonal Antibody
Catalog # USBI136350
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Prostaglandin Dehydrogenase 15
  • Antigen symbol:
    Prostaglandin Dehydrogenase 15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Bovine,
    Guinea pig
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    92 to 105
  • Storage buffer:
    TBSpH 7,4, 0,5mg/ml BSA, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:200; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: Prostaglandin Dehydrogenase 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Guinea pig