Anti-Mucin 16 Rabbit Polyclonal Antibody
Catalog # USBI136271
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mucin 16
- Antigen symbol:MUC16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 150mM sodium chloride, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:100-1:500; ELISA: 1:20000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Mucin 16
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human