Anti-APAF1 Rabbit Polyclonal Antibody
Catalog # USBI135900
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Apoptotic peptidase activating factor 1
- Antigen symbol:APAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Antigen synonyms:CED4
- Storage buffer:PBS,(without Mg2+, Ca2+), pH 7,4, 150mM sodium chloride, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: APAF1 (apoptotic peptidase activating factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human